missing translation for 'onlineSavingsMsg'
Learn More

1-Aza-15-crown-5, 97%

Catalog No. AAH51064MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51064MD 250 mg
AAH5106403 1 g
AAH5106406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH51064MD Supplier Thermo Scientific Chemicals Supplier No. H51064MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 66943-05-3 | C10H21NO4 | 219.281 g/mol

1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.

Solubility
Soluble in water and methanol.

Notes
Air sensitive. Store away from oxidizing agents.

Chemical Identifiers

CAS 66943-05-3
Molecular Formula C10H21NO4
Molecular Weight (g/mol) 219.281
MDL Number MFCD00075465
InChI Key BJUOQSZSDIHZNP-UHFFFAOYSA-N
Synonym aza-15-crown-5, 1-aza-15-crown 5-ether, 1-aza-15-crown-5, 1-aza-15-crown, 1-aza-4,7,10,13-tetraoxacyclopentadecane
PubChem CID 544820
IUPAC Name 1,4,7,10-tetraoxa-13-azacyclopentadecane
SMILES C1COCCOCCOCCOCCN1

Specifications

Quantity 250 mg
Beilstein 971959
Sensitivity Air Sensitive
Solubility Information Soluble in water and methanol.
Formula Weight 219.28
Percent Purity 97%
Chemical Name or Material 1-Aza-15-crown-5
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 266-523-3
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only