accessibility menu, dialog, popup

UserName

CH3CH3(S)HNO(R)O(R)HOOONHNNH2NH2

CAS RN 66701-25-5

IUPAC Name: (2R,3R)-3-{[(1S)-1-({4-[(diaminomethylidene)amino]butyl}carbamoyl)-3-methylbutyl]carbamoyl}oxirane-2-carboxylic acid
Synonyms: (2R,3R)-3-{[(1S)-1-({4-[(diaminomethylidene)amino]butyl}carbamoyl)-3-methylbutyl]carbamoyl}oxirane-2-carboxylic acid
Molecular Weight (g/mol): 357.41
Molecular Formula: C15H27N5O5
InChi Key: LTLYEAJONXGNFG-HBNTYKKESA-N
SMILES: CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(O)=O)C(=O)NCCCCN=C(N)N
Quantity 
  • (1)
  • (1)
  • (2)
Form 
  • (2)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results