missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ E-64 Protease Inhibitor

Catalog No. 3248905MG
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3248905MG 5 mg
3248901MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3248905MG Supplier MilliporeSigma™ Supplier No. 3248905MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The E-64 Protease Inhibitor, also referenced under CAS 66701-25-5, controls the biological activity of E-64 Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Irreversible inhibitor of cysteine proteases. Interacts with the Sn subsites of proteases. Has no action on cysteine residues in other proteins. Inhibits activation-induced programmed cell death and restores defective immune responses in HIV+ donors. Specific active site titrant.

Specifications

Color White
Product Type Inhibitor
Target Proteins cysteine proteases
CAS 66701-25-5
Molecular Formula C15H27N5O5
Formula Weight 357.4g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Purity 98% by HPLC