accessibility menu, dialog, popup

UserName

CAS RN 66-77-3

IUPAC Name: naphthalene-1-carbaldehyde
Synonyms: 1-naphthaldehyde alphaα-naphthaldehyde α-naphthal
Molecular Weight (g/mol): 156.18
Molecular Formula: C11H8O
InChi Key: SQAINHDHICKHLX-UHFFFAOYSA-N
SMILES: O=CC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (3)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)