missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaldehyde, 97%

Catalog No. AAA1561030
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1561030 250 g
AAA1561018 50 g
AAA156100B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1561030 Supplier Thermo Scientific Chemicals Supplier No. A1561030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 66-77-3 | C11H8O | 156.18 g/mol

1-Naphthaldehyde is used in the synthesis of single-crystalline homochiral porous metal-organic frameworks (MOFs). It is also a starting material for the Canizzaro reaction to produce 1-Naphthoic acid and 1-Naphthalenemethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthaldehyde is used in the synthesis of single-crystalline homochiral porous metal-organic frameworks (MOFs). It is also a starting material for the Canizzaro reaction to produce 1-Naphthoic acid and 1-Naphthalenemethanol.

Solubility
Soluble in ethanol, ether, acetone, benzene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agent, air.

Chemical Identifiers

CAS 66-77-3
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.18
MDL Number MFCD00004003
InChI Key SQAINHDHICKHLX-UHFFFAOYSA-N
Synonym 1-naphthaldehyde, 1-formylnaphthalene, 1-naphthalenecarboxaldehyde, 1-naphthylaldehyde, alpha-naphthal, alpha-naphthaldehyde, naphthaldehyde, .alpha.-naphthal, alpha-naphthylcarboxaldehyde, alpha-naphthylaldehyde
PubChem CID 6195
ChEBI CHEBI:52367
IUPAC Name naphthalene-1-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 1°C to 2°C
Density 1.148
Boiling Point 160°C to 161°C (15 mmHg)
Flash Point 112°C (233°F)
Odor Pungent
Refractive Index 1.653
Quantity 250 g
Beilstein 386082
Sensitivity Air sensitive; May form a precipitate
Solubility Information Soluble in ethanol,ether,acetone,benzene. Insoluble in water.
Formula Weight 156.18
Percent Purity 97%
Chemical Name or Material 1-Naphthaldehyde
Show More Show Less
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 200-633-4
RTECSNumber QJ0190000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only