missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaldehyde 95.0+%, TCI America™

Catalog No. N0002500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0002500ML 500 mL
N000225ML 25 mL
N0002100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0002500ML Supplier TCI America Supplier No. N0002500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66-77-3
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.18
MDL Number MFCD00004003
InChI Key SQAINHDHICKHLX-UHFFFAOYSA-N
Synonym 1-naphthaldehyde, 1-formylnaphthalene, 1-naphthalenecarboxaldehyde, 1-naphthylaldehyde, alpha-naphthal, alpha-naphthaldehyde, naphthaldehyde, .alpha.-naphthal, alpha-naphthylcarboxaldehyde, alpha-naphthylaldehyde
PubChem CID 6195
ChEBI CHEBI:52367
IUPAC Name naphthalene-1-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 2°C
Color Yellow
Boiling Point 291°C
Quantity 500 mL
Formula Weight 156.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Naphthaldehyde
EINECSNumber (4)-1431
RTECSNumber QJ0190000
TSCA Yes