accessibility menu, dialog, popup

UserName

CAS RN 657-84-1

IUPAC Name: sodium 4-methylbenzene-1-sulfonate
Synonyms: sodium p-tolyl sulfonate
Molecular Weight (g/mol): 194.18
Molecular Formula: C7H7NaO3S
InChi Key: KVCGISUBCHHTDD-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)