accessibility menu, dialog, popup

UserName

OO

CAS RN 646-06-0

IUPAC Name: 1,3-dioxolane
Synonyms: dioxolane 1,3-dioxalane
Molecular Weight (g/mol): 74.08
Molecular Formula: C3H6O2
InChi Key: WNXJIVFYUVYPPR-UHFFFAOYSA-N
SMILES: C1COCO1
Boiling Point 
  • (4)
  • (2)
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (11)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (4)
  • (2)
  • (8)
  • (4)
  • (2)
  • (3)
  • (5)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (11)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)
  • (3)
  • (4)