missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99.5%, stab.

Catalog No. AA42897M1
Change view
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42897M1 4 x 1 L
AA42897AK 250 mL
AA42897K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42897M1 Supplier Thermo Scientific Chemicals Supplier No. 042897M1
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

As a solvent, in Mannich reaction, as an electrolyte in batteries | CAS: 646-06-0 | C3H6O2 | 74.079 g/mol

As a solvent, in Mannich reaction, as an electrolyte in batteries.1,3-Dioxolane is used as a solvent for paints, cellulose esters and in organic synthesis. It is used as an extragent for waxes, fats and oils, stabilizer for halogenated hydrocarbons and co monomer in polyacetals preparation. It is used as finishing agent and swelling agent in textiles. It acts as an intermediate for the preparation of acyclovir and vandetanib. It is an essential ingredient in industrial polymers. In polymerization reactions, it acts as a chain length regulator and a chain transfer agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 646-06-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 12586
ChEBI CHEBI:87597
IUPAC Name 1,3-dioxolane
SMILES C1COCO1

Specifications

Melting Point -95°C
Density 1.06 g/mL
Boiling Point 74°C to 75°C
Flash Point 1°C (34°F)
Refractive Index 1.4
Quantity 4 x 1 L
UN Number UN1166
Solubility Information Miscible with water,ethanol,ether,n-octanol,acetone,chloroform,dichloromethane,ethyl Acetate and methanol.
Formula Weight 74.08
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material 1,3-Dioxolane, Stabilized
Show More Show Less
Hazard Category H225-H319
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIOXOLANE
EINECSNumber 211-463-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only