missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane (stabilized with BHT) 98.0+%, TCI America™

Catalog No. D5539500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5539500ML 500 mL
D553925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5539500ML Supplier TCI America Supplier No. D5539500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 646-06-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 12586
ChEBI CHEBI:87597
IUPAC Name 1,3-dioxolane
SMILES C1COCO1

Specifications

Color Yellow
Boiling Point 74°C
Quantity 500 mL
UN Number 1166
Formula Weight 74.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dioxolane (stabilized with BHT)
EINECSNumber (5)-0500
RTECSNumber JH6760000
TSCA Yes