missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99.5+%, pure, stabilized

Catalog No. AC299480250
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC299480250 25 L Metal drum
AC299480010 1 L Glass Bottle
AC299480025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC299480250 Supplier Thermo Scientific Chemicals Supplier No. 299480250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 646-06-0 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 646-06-0 , 128-37-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -95°C
Color White
Density 1.0600g/mL
Boiling Point 74.0°C to 75.0°C
Flash Point −6°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Metal drum
Refractive Index 1.3990 to 1.4010
Quantity 25 L
Beilstein 19, 2
Fieser 14,281
Specific Gravity 1.06
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and acetone
Stabilizer 75ppm BHT max.
Viscosity 0.66 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 99.5+%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Dioxolane, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
Wash f
EINECSNumber 211-463-5

RUO – Research Use Only