missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99%, Spectrum™ Chemical

Catalog No. 18608060
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608060 500 mL Steel Can
18608059 4 L Steel Can
18608057 20 L Steel Can
18608058 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608060 Supplier Spectrum Chemical Mfg Cor Supplier No. D2371500ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 646-06-0

Specifications

Acidity 100 ppm
CAS Max % 1
Boiling Point 74 to 78°C
Packaging Steel Can
Quantity 500 mL
Percent Purity 0.99
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.