accessibility menu, dialog, popup

UserName

HHClClCH3H3CH2NCH3NH2CH3

CAS RN 64285-73-0

IUPAC Name: dihydrogen 3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine dichloride
Synonyms: dihydrogen tetramethylbenzidine dichloride
Molecular Weight (g/mol): 313.27
Molecular Formula: C16H22Cl2N2
InChi Key: NYNRGZULARUZCC-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results