missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine dihydrochloride, 99%

Catalog No. AAH60245MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60245MD 250 mg
AAH6024503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60245MD Supplier Thermo Scientific Chemicals Supplier No. H60245MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64285-73-0 | C16H22Cl2N2 | 313.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64285-73-0
Molecular Formula C16H22Cl2N2
Molecular Weight (g/mol) 313.27
MDL Number MFCD00012961
InChI Key NYNRGZULARUZCC-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine dihydrochloride, tmb dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethyl-, dihydrochloride, tetramethylbenzidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethyl-, hydrochloride 1:2, 4-4-amino-3,5-dimethylphenyl-2,6-dimethylaniline dihydrochloride, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, 3,3',5,5'-tetramethyl 1,1'-biphenyl-4,4'-diamine dihydrochloride
PubChem CID 174033
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1

Specifications

Melting Point 300°C
Quantity 250 mg
Sensitivity Light and moisture sensitive
Formula Weight 313.27
Percent Purity 99%
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine dihydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 264-769-6
TSCA Yes
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only