accessibility menu, dialog, popup

UserName

H3CI

CAS RN 638-45-9

IUPAC Name: 1-iodohexane
Synonyms: 1-iodohexane
Molecular Weight (g/mol): 212.07
Molecular Formula: C6H13I
InChi Key: ANOOTOPTCJRUPK-UHFFFAOYSA-N
SMILES: CCCCCCI
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)