missing translation for 'onlineSavingsMsg'
Learn More

1-Iodohexane (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. I005925G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I005925G 25 g
I0059100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I005925G Supplier TCI America Supplier No. I005925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 638-45-9
Molecular Formula C6H13I
Molecular Weight (g/mol) 212.074
MDL Number MFCD00001102
InChI Key ANOOTOPTCJRUPK-UHFFFAOYSA-N
PubChem CID 12527
IUPAC Name 1-iodohexane
SMILES CCCCCCI

Specifications

Color Yellow
Boiling Point 180°C
Quantity 25 g
UN Number 3082
Synonym hexyl iodide, n-hexyl iodide, hexane, 1-iodo, 1-hexyl iodide, iodohexane, 1-iodo-hexane, hexane,1-iodo, n-hexyliodide, iodo-hexane, 1-iodanylhexane
Formula Weight 212.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodohexane (stabilized with Copper chip)