missing translation for 'onlineSavingsMsg'
Learn More

1-Iodohexane, 98%, stab. with copper

Catalog No. AAA1378436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1378436 500 g
AAA1378414 25 g
AAA1378422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1378436 Supplier Thermo Scientific Chemicals Supplier No. A1378436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 638-45-9 | C6H13I | 212.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 638-45-9
Molecular Formula C6H13I
Molecular Weight (g/mol) 212.074
MDL Number MFCD00001102
InChI Key ANOOTOPTCJRUPK-UHFFFAOYSA-N
Synonym hexyl iodide, n-hexyl iodide, hexane, 1-iodo, 1-hexyl iodide, iodohexane, 1-iodo-hexane, hexane,1-iodo, n-hexyliodide, iodo-hexane, 1-iodanylhexane
PubChem CID 12527
IUPAC Name 1-iodohexane
SMILES CCCCCCI

Specifications

Melting Point -75°C
Density 1.437
Boiling Point 179°C to 181°C
Flash Point 61°C (141°F)
Refractive Index 1.492
Quantity 500 g
Beilstein 505971
Sensitivity Light sensitive
Formula Weight 212.08
Percent Purity 98%
Chemical Name or Material 1-Iodohexane, stabilized with copper
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-339-0
RTECSNumber MO3855000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only