accessibility menu, dialog, popup

UserName

CAS RN 637-32-1

IUPAC Name: hydrogen 1-({amino[(4-chlorophenyl)amino]methylidene}amino)-N'-(propan-2-yl)methanimidamide chloride
Synonyms: hydrogen proguanil chloride
Molecular Weight (g/mol): 290.19
Molecular Formula: C11H17Cl2N5
InChi Key: SARMGXPVOFNNNG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)N=C(N)N=C(N)NC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)