missing translation for 'onlineSavingsMsg'
Learn More

Proguanil hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018861
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018861 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018861 Supplier Merck Emd Millipore Supplier No. PHR17131G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Proguanil hydrochloride is a biguanide compound, widely used as an antimalarial drug. It exhibits sporontocidal activity, thereby rendering the gametocytes non-infective to the mosquito vector.Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 637-32-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C11H16 ClN5·HCl
MDL Number MFCD01732193
Synonym Proguanil hydrochloride; Chlorguanide; N1-(4-Chlorophenyl)-N5-isopropylbiguanide
Type Certified Reference Material
Quantity 1 g
Formula Weight 290.19
Grade Certified Reference Material