missing translation for 'onlineSavingsMsg'
Learn More

Proguanil hydrochloride, 97%

Catalog No. AC461431000
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461431000 100 mg
AC461430250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461431000 Supplier Thermo Scientific Chemicals Supplier No. 461431000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 637-32-1 | C11H16ClN5·HCl | 290.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 637-32-1
Molecular Formula C11H16ClN5·HCl
Molecular Weight (g/mol) 290.19
InChI Key SARMGXPVOFNNNG-UHFFFAOYSA-N
Synonym proguanil hydrochloride, diguanyl, paludrine, proguanil hcl, chlorguanide hydrochloride, chloroquanil, bigumalum, chloroguanide hydrochloride, tirian hydrochloride, palusil hydrochloride
PubChem CID 9570076
IUPAC Name (1E)-1-[amino-(4-chloroanilino)methylidene]-2-propan-2-ylguanidine;hydrochloride
SMILES CC(C)N=C(N)N=C(N)NC1=CC=C(C=C1)Cl.Cl

Specifications

Melting Point 245.0°C to 250.0°C
Quantity 100 mg
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (1 g, 105°C)
Assay Percent Range 96% min. (HPLC)
Formula Weight 290.19
Percent Purity 97%
Chemical Name or Material Proguanil hydrochloride
Hazard Category Acute toxicity (category 3)
Hazard Statement Toxic if swallowed
EINECSNumber 211-283-7

RUO – Research Use Only