accessibility menu, dialog, popup

UserName

NH2NCl

CAS RN 6298-19-7

IUPAC Name: 2-chloropyridin-3-amine
Synonyms: 2-chloropyridin-3-amine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: MEQBJJUWDCYIAB-UHFFFAOYSA-N
SMILES: NC1=CC=CN=C1Cl
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)