missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-chloropyridine 98.0+%, TCI America™

Catalog No. A083625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A083625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A083625G Supplier TCI America Supplier No. A083625G

Chemical Identifiers

CAS 6298-19-7
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00006238
InChI Key MEQBJJUWDCYIAB-UHFFFAOYSA-N
Synonym 3-amino-2-chloropyridine, 2-chloro-3-aminopyridine, 2-chloro-3-pyridinamine, 3-pyridinamine, 2-chloro, 2-chloro-3-amino pyridine, 2-chloro-3-pyridylamine, 2-chloro-3-amino-pyridine, 2-chloro-pyridin-3-ylamine, 2-chloro-3-pyridinylamine, 3-amino-2-chloro-pyridine
PubChem CID 80528
IUPAC Name 2-chloropyridin-3-amine
SMILES NC1=CC=CN=C1Cl

Specifications

Melting Point 78°C
Boiling Point 135°C
Quantity 25 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-chloropyridine