missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-chloropyridine, 98+%

Catalog No. AAA1583030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1583030 250 g
AAA1583009 10 g
AAA1583018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1583030 Supplier Thermo Scientific Chemicals Supplier No. A1583030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6298-19-7 | C5H5ClN2 | 128.56 g/mol

It is an intermediate of medicine and agricultural chemicals. It is involved in the Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate of medicine and agricultural chemicals. It is involved in the Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.

Solubility
Soluble in methanol and water. (30 g/L).

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 6298-19-7
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.56
MDL Number MFCD00006238
InChI Key MEQBJJUWDCYIAB-UHFFFAOYSA-N
Synonym 3-amino-2-chloropyridine, 2-chloro-3-aminopyridine, 2-chloro-3-pyridinamine, 3-pyridinamine, 2-chloro, 2-chloro-3-amino pyridine, 2-chloro-3-pyridylamine, 2-chloro-3-amino-pyridine, 2-chloro-pyridin-3-ylamine, 2-chloro-3-pyridinylamine, 3-amino-2-chloro-pyridine
PubChem CID 80528
IUPAC Name 2-chloropyridin-3-amine
SMILES NC1=CC=CN=C1Cl

Specifications

Melting Point 76°C to 80°C
Boiling Point 130°C to 134°C (22 mmHg)
Flash Point 185°C (365°F)
Odor Unpleasant
Quantity 250 g
Beilstein 109814
Solubility Information Soluble in methanol and water. (30g/L).
Formula Weight 128.56
Percent Purity ≥98%
Chemical Name or Material 3-Amino-2-chloropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-572-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only