CAS RN 6284-79-3
IUPAC Name:
(3,4-dichlorophenyl)(phenyl)methanone
Synonyms:
(3,4-dichlorophenyl)(phenyl)methanone
Molecular Weight (g/mol):
251.11
Molecular Formula:
C13H8Cl2O
InChi Key:
LLUPHTAYNHAVQT-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1Cl)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
3,4-Dichlorobenzophenone 98.0+%, TCI America™
CAS: 6284-79-3 Molecular Formula: C13H8Cl2O Molecular Weight (g/mol): 251.106 MDL Number: MFCD00000552 InChI Key: LLUPHTAYNHAVQT-UHFFFAOYSA-N Synonym: 3,4-dichlorobenzophenone,3,4-dichlorophenyl phenyl methanone,benzophenone, 3,4-dichloro,methanone, 3,4-dichlorophenyl phenyl,3,4-dichlorophenyl phenyl ketone,pubchem7488,benzophenone,4-dichloro,methanone,4-dichlorophenyl phenyl,3,4-dichlorophenyl-phenylmethanone,3,4-dichlorophenyl phenyl-methanone PubChem CID: 80494 IUPAC Name: (3,4-dichlorophenyl)-phenylmethanone SMILES: C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)Cl)Cl