accessibility menu, dialog, popup

UserName

CAS RN 6279-86-3

IUPAC Name: triethyl methanetricarboxylate
Synonyms: triethyl methanetricarboxylate
Molecular Weight (g/mol): 232.23
Molecular Formula: C10H16O6
InChi Key: AGZPNUZBDCYTBB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C(=O)OCC)C(=O)OCC
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)