missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Methanetricarboxylate 97.0+%, TCI America™

Catalog No. T14815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14815G 5 g
T148125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14815G Supplier TCI America Supplier No. T14815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6279-86-3
Molecular Formula C10H16O6
Molecular Weight (g/mol) 232.232
MDL Number MFCD00009150
InChI Key AGZPNUZBDCYTBB-UHFFFAOYSA-N
Synonym tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
PubChem CID 80471
IUPAC Name triethyl methanetricarboxylate
SMILES CCOC(=O)C(C(=O)OCC)C(=O)OCC

Specifications

Color White
Boiling Point 253°C
Quantity 5 g
Formula Weight 232.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Triethyl Methanetricarboxylate