CAS RN 62625-28-9
Bromophenol Blue Solution, 0.04%/Certified, Fisher Chemical
Thermo Scientific Chemicals Bromophenol Blue, sodium salt, pure, water soluble, indicator
CAS: 62625-28-9 Molecular Formula: C19H9Br4NaO5S Molecular Weight (g/mol): 691.942 MDL Number: MFCD00013793 InChI Key: TWKDJMRHSLWRFI-UHFFFAOYSA-M Synonym: 3',3'',5',5''-Tetrabromophenolsulfonephthalein PubChem CID: 3791690 IUPAC Name: sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate SMILES: C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
Thermo Scientific Chemicals Bromophenol Blue, sodium salt, pure, water soluble, indicator
CAS: 62625-28-9 Molecular Formula: C19H9Br4NaO5S Molecular Weight (g/mol): 691.942 MDL Number: MFCD00013793 InChI Key: TWKDJMRHSLWRFI-UHFFFAOYSA-M Synonym: 3',3'',5',5''-Tetrabromophenolsulfonephthalein PubChem CID: 3791690 IUPAC Name: sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate SMILES: C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
Bromophenol Blue Sodium Salt, ACS Reagent Grade, Ricca Chemical
CAS: 62625-28-9 Molecular Formula: C19H9Br4NaO5S Molecular Weight (g/mol): 691.942 InChI Key: TWKDJMRHSLWRFI-UHFFFAOYSA-M PubChem CID: 3791690 IUPAC Name: sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate SMILES: C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
Bromophenol Blue, 0.04% Aqueous, pH 3.0 to 4.6 Yellow to Blue, Certified, LabChem™
CAS: 62625-28-9 Molecular Formula: C19H9Br4NaO5S Molecular Weight (g/mol): 691.942 InChI Key: TWKDJMRHSLWRFI-UHFFFAOYSA-M PubChem CID: 3791690 IUPAC Name: sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate SMILES: C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
Bromophenol Blue Sodium Salt, ACS Grade, LabChem™
CAS: 62625-28-9 Molecular Formula: C19H9Br4NaO5S Molecular Weight (g/mol): 691.942 InChI Key: TWKDJMRHSLWRFI-UHFFFAOYSA-M PubChem CID: 3791690 IUPAC Name: sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate SMILES: C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
Bromophenol Blue, 0.1% Aqueous, pH 3.0 to 4.6 Yellow to Blue, Certified, LabChem™
CAS: 62625-28-9 Molecular Formula: C19H9Br4NaO5S Molecular Weight (g/mol): 691.942 InChI Key: TWKDJMRHSLWRFI-UHFFFAOYSA-M PubChem CID: 3791690 IUPAC Name: sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate SMILES: C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
Bromophenol Blue, Sodium Salt, MP Biomedicals™
Synonym: 3',3”,5',5”-tetrabromophenolsulfonphthalein sodium salt,3', 3”, 5'
Bromophenol Blue TS, (U.S.P. Test Solution), Spectrum™ Chemical
CAS: 62625-28-9 Molecular Formula: C19H9Br4NaO5S Molecular Weight (g/mol): 691.94 InChI Key: TWKDJMRHSLWRFI-UHFFFAOYSA-M IUPAC Name: sodium 2-[(3,5-dibromo-4-hydroxyphenyl)(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzene-1-sulfonate SMILES: [Na+].OC1=C(Br)C=C(C=C1Br)C(=C1C=C(Br)C(=O)C(Br)=C1)C1=CC=CC=C1S([O-])(=O)=O