missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue Indicator, 0.4% (w/v), Ricca Chemical

Catalog No. 136016
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
136016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 136016 Supplier Ricca Chemical Company Supplier No. 136016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Bromophenol Blue Indicator
CAS 62625-28-9, 7732-18-5
CAS Min % 98.0, 0.39
CAS Max % 100.0, 0.41
Concentration 0.4% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Dark Reddish Purple
pH 4
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.4% W/V
Grade Indicator
Show More Show Less