missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, Sodium Salt, MP Biomedicals™

Catalog No. ICN15250605
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15250605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15250605 Supplier MP Biomedicals Inc Supplier No. 0215250605

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bromophenol Blue, Sodium Salt is a tracking dye.

  • Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.
  • It is a tracking dye for alkaline and neutral buffer systems, as well as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.

Chemical Identifiers

Synonym 3',3”,5',5”-tetrabromophenolsulfonphthalein sodium salt, 3', 3”, 5'

Specifications

CAS 62625-28-9
Molecular Formula C19H9Br4NaO5S
Color Green
Quantity 5 g
InChI Key TWKDJMRHSLWRFI-UHFFFAOYSA-M
Physical Form Crystalline Powder
SMILES C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
Molecular Weight (g/mol) 691.942
PubChem CID 3791690
Formula Weight 692
Chemical Name or Material Bromophenol blue, sodium salt, For ACS analysis
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.