accessibility menu, dialog, popup

UserName

CAS RN 624-73-7

IUPAC Name: 1,2-diiodoethane
Synonyms: 1,2-diiodoethane
Molecular Weight (g/mol): 281.86
Molecular Formula: C2H4I2
InChi Key: GBBZLMLLFVFKJM-UHFFFAOYSA-N
SMILES: ICCI
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)