missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodoethane, 98%

Catalog No. AAL0301914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0301914 25 g
AAL0301906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0301914 Supplier Thermo Scientific Chemicals Supplier No. L0301914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 624-73-7 | C2H4I2 | 281.863 g/mol

1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diiodoethane is used as C-X bond formation reagent. It is also used as Pharmaceutical intermediates.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store at 4°C. Air & Light Sensitive. Incompatible with strong bases, strong oxidizing agents, heat and light. Protect from heat.

Chemical Identifiers

CAS 624-73-7
Molecular Formula C2H4I2
Molecular Weight (g/mol) 281.863
MDL Number MFCD00001093
InChI Key GBBZLMLLFVFKJM-UHFFFAOYSA-N
Synonym ethane, 1,2-diiodo, ethylene diiodide, ethylene iodide, 1,2-diiodethane, 1,2-diiodo ethane, 1,2-bis iodanyl ethane, ch2ich2i, 1,2-diiodoethane, acmc-1b2u1, ksc358c5b
PubChem CID 12224
IUPAC Name 1,2-diiodoethane
SMILES C(CI)I

Specifications

Melting Point 80°C to 84°C
Density 2.13
Quantity 25 g
Beilstein 1730870
Sensitivity Light sensitive
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 281.86
Percent Purity 98%
Chemical Name or Material 1,2-Diiodoethane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-859-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only