accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 624-24-8

IUPAC Name: methyl pentanoate
Synonyms: methyl valerate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: HNBDRPTVWVGKBR-UHFFFAOYSA-N
SMILES: CCCCC(=O)OC
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)