missing translation for 'onlineSavingsMsg'
Learn More

Methyl valerate, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020191
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020191 1 mL
111019392 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020191 Supplier Merck Emd Millipore Supplier No. 945601ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methyl valerate is a methyl ester, which may find application as a surrogate component in the production of biodiesel.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 624-24-8
Density 0.875 g/mL (at 25°C)
Flash Point 22°C
Physical Form Neat
Linear Formula CH3(CH2)3 CO2 CH3
MDL Number MFCD00009478
Boiling Point 128°C
Synonym Methyl pentanoate
Type Analytical Standard
Quantity 1 mL
Formula Weight 116.16
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less