missing translation for 'onlineSavingsMsg'
Learn More

Methyl Valerate 99.5+%, TCI America™

Catalog No. S03035ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03035ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03035ML Supplier TCI America Supplier No. S03035ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 624-24-8
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009478
InChI Key HNBDRPTVWVGKBR-UHFFFAOYSA-N
Synonym methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn
PubChem CID 12206
IUPAC Name methyl pentanoate
SMILES CCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 128°C
Quantity 5 mL
UN Number 3272
Formula Weight 116.16
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Valerate [Standard Material for GC]
EINECSNumber (2)-0776
RTECSNumber YV7750500
TSCA Yes