missing translation for 'onlineSavingsMsg'
Learn More

Methyl Valerate 99.0+%, TCI America™

Catalog No. V000525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V000525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V000525ML Supplier TCI America Supplier No. V000525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 624-24-8
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009478
InChI Key HNBDRPTVWVGKBR-UHFFFAOYSA-N
Synonym methyl valerate, pentanoic acid, methyl ester, methyl n-valerate, methyl valerianate, valeric acid, methyl ester, valeric acid methyl ester, unii-zw21jjj9vn, pentanoic acid methyl ester, fema no. 2752, zw21jjj9vn
PubChem CID 12206
IUPAC Name methyl pentanoate
SMILES CCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 128°C
Quantity 25 mL
UN Number 3272
Formula Weight 116.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Valerate
EINECSNumber (2)-0776
RTECSNumber YV7750500
TSCA Yes