accessibility menu, dialog, popup

UserName

CH3SiCH3CH3OH2C

CAS RN 6213-94-1

IUPAC Name: (ethenyloxy)trimethylsilane
Synonyms: (ethenyloxy)trimethylsilane
Molecular Weight (g/mol): 116.24
Molecular Formula: C5H12OSi
InChi Key: HFTNNOZFRQLFQB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC=C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (4)