missing translation for 'onlineSavingsMsg'
Learn More

Vinyloxytrimethylsilane, 97%

Catalog No. AAH2687606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2687606 5 g
AAH2687603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2687606 Supplier Thermo Scientific Chemicals Supplier No. H2687606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6213-94-1 | C5H12OSi | 116.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6213-94-1
Molecular Formula C5H12OSi
Molecular Weight (g/mol) 116.235
MDL Number MFCD00054764
InChI Key HFTNNOZFRQLFQB-UHFFFAOYSA-N
Synonym trimethyl vinyloxy silane, vinyloxytrimethylsilane, silane, ethenyloxy trimethyl, tmso-ethene, vinyloxy-trimethylsilane, trimethylsiloxy ethylene, snqhhgciiezjp@, acmc-209mzx, ethenoxy trimethyl silane, trimethylsilyl vinyl ether
PubChem CID 80344
IUPAC Name ethenoxy(trimethyl)silane
SMILES C[Si](C)(C)OC=C

Specifications

Boiling Point 74°C
Quantity 5 g
Flash Point −19°C (−2°F)
Refractive Index 1.389
Sensitivity Moisture sensitive
Density 0.779
UN Number UN1993
Formula Weight 116.24
Percent Purity 97%
Chemical Name or Material Vinyloxytrimethylsilane
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 228-281-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only