CAS RN 621-88-5
IUPAC Name:
2-phenoxyacetamide
Synonyms:
phenoxyacetamide
Molecular Weight (g/mol):
151.17
Molecular Formula:
C8H9NO2
InChi Key:
AOPRXJXHLWYPQR-UHFFFAOYSA-N
SMILES:
NC(=O)COC1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-Phenoxyacetamide, 98%
2-Phenoxyacetamide, 98%
CAS: 621-88-5 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.17 MDL Number: MFCD00017140 InChI Key: AOPRXJXHLWYPQR-UHFFFAOYSA-N Synonym: phenoxyacetamide,acetamide, 2-phenoxy,phenoxyacetic amide,alpha-phenoxyacetamide,2-phenoxyethanamide,2-phenoxy acetamide,pubchem12494,acmc-20ajx9,maybridge1_008727,po6 PubChem CID: 69314 IUPAC Name: 2-phenoxyacetamide SMILES: NC(=O)COC1=CC=CC=C1