missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyacetamide, 98%

Catalog No. AAA1493414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1493414 25 g
AAA1493406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1493414 Supplier Thermo Scientific Chemicals Supplier No. A1493414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 621-88-5 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-88-5
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00017140
InChI Key AOPRXJXHLWYPQR-UHFFFAOYSA-N
Synonym phenoxyacetamide, acetamide, 2-phenoxy, phenoxyacetic amide, alpha-phenoxyacetamide, 2-phenoxyethanamide, 2-phenoxy acetamide, pubchem12494, acmc-20ajx9, maybridge1_008727, po6
PubChem CID 69314
IUPAC Name 2-phenoxyacetamide
SMILES NC(=O)COC1=CC=CC=C1

Specifications

Melting Point 100°C to 104°C
Quantity 25 g
Beilstein 1941781
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material 2-Phenoxyacetamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-713-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only