accessibility menu, dialog, popup

UserName

H2NNHOHO

CAS RN 619-67-0

IUPAC Name: 4-hydrazinylbenzoic acid
Synonyms: 4-hydrazinylbenzoic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: PCNFLKVWBDNNOW-UHFFFAOYSA-N
SMILES: NNC1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (6)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)