missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic acid, 98%

Catalog No. AC411791000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411791000 100 g Glass bottle
AC411790050 5 g Glass bottle
AC411790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC411791000 Supplier Thermo Scientific Chemicals Supplier No. 411791000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 619-67-0 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-67-0
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007581
InChI Key PCNFLKVWBDNNOW-UHFFFAOYSA-N
Synonym 4-hydrazinobenzoic acid, p-hydrazinobenzoic acid, benzoic acid, 4-hydrazino, p-carboxyphenylhydrazine, 4-carboxyphenyl hydrazine, 4-carboxyphenylhydrazine, 4-hydrazino benzoic acid, benzoic acid, p-hydrazino, unii-b1r368je7o, ccris 7911
PubChem CID 12089
IUPAC Name 4-hydrazinylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)NN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: < 10g/L (20°C). Other solubilities: limited soluble in alcohols
Formula Weight 152.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydrazinobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-609-5

RUO – Research Use Only