missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic acid, 97%

Catalog No. AAA1387430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1387430 250 g
AAA1387409 10 g
AAA1387418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1387430 Supplier Thermo Scientific Chemicals Supplier No. A1387430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 619-67-0 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-67-0
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007581
InChI Key PCNFLKVWBDNNOW-UHFFFAOYSA-N
Synonym 4-hydrazinobenzoic acid, p-hydrazinobenzoic acid, benzoic acid, 4-hydrazino, p-carboxyphenylhydrazine, 4-carboxyphenyl hydrazine, 4-carboxyphenylhydrazine, 4-hydrazino benzoic acid, benzoic acid, p-hydrazino, unii-b1r368je7o, ccris 7911
PubChem CID 12089
IUPAC Name 4-hydrazinylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)NN

Specifications

Melting Point ∼216°C (decomposition)
Quantity 250 g
Beilstein 387378
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material 4-Hydrazinobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-609-5
RTECSNumber DH1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only