CAS RN 61825-94-3
IUPAC Name:
(1R,2R)-cyclohexane-1,2-diamine; 1,3-dioxa-2-platinacyclopentane-4,5-dione
Synonyms:
(1R,2R)-cyclohexane-1,2-diamine; 1,3-dioxa-2-platinacyclopentane-4,5-dione
Molecular Weight (g/mol):
397.29
Molecular Formula:
C8H14N2O4Pt
InChi Key:
ZROHGHOFXNOHSO-WHMNLBETNA-L
SMILES:
O=C1O[Pt]OC1=O.N[C@@H]1CCCC[C@H]1N
1
–
4
of
4
results
2
Thermo Scientific Chemicals Oxaliplatin
CAS: 61825-94-3 Molecular Formula: C8H14N2O4Pt Synonym: NSC 266046;
3
Promotions Available
Thermo Scientific Chemicals Oxaliplatin, 99%
Thermo Scientific Chemicals Oxaliplatin, 99%
CAS: 61825-94-3 Molecular Formula: C8H14N2O4Pt Molecular Weight (g/mol): 397.29 MDL Number: MFCD00866327 InChI Key: ZROHGHOFXNOHSO-WHMNLBETNA-L IUPAC Name: (1R,2R)-cyclohexane-1,2-diamine; 1,3-dioxa-2-platinacyclopentane-4,5-dione SMILES: O=C1O[Pt]OC1=O.N[C@@H]1CCCC[C@H]1N
4
Oxaliplatin, TCI America™
CAS: 61825-94-3 Molecular Formula: C8H14N2O4Pt Molecular Weight (g/mol): 397.29 MDL Number: MFCD00866327 InChI Key: ZROHGHOFXNOHSO-WHMNLBETNA-L Synonym: (trans-1,2-Cyclohexanediamine)oxalatoplatinum(II) IUPAC Name: (1R,2R)-cyclohexane-1,2-diamine; 1,3-dioxa-2-platinacyclopentane-4,5-dione SMILES: O=C1O[Pt]OC1=O.N[C@@H]1CCCC[C@H]1N