missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxaliplatin

Catalog No. AAJ66586MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66586MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66586MF Supplier Thermo Scientific Chemicals Supplier No. J66586MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61825-94-3 | C8H14N2O4Pt, C8H14N2O4Pt | 397.29 g/mol

Oxaliplatin is an anti-tumor agent with activity against colorectal cancer. It shows antitumor activity in cell lines with acquired cisplatin resistance. It is used to treat cancer of the gullet (oesophagus). It is used as adjuvant therapy in patients who have had surgery to remove the cancer. It is used to treat testicular cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61825-94-3
Molecular Formula C8H14N2O4Pt
Synonym NSC 266046;

Specifications

Quantity 50 mg
Description White to off-white
Content And Storage -30°C to -10°C
Formula Weight 397.29
Solubility Soluble in water with heating and/or sonication
Product Type Oxaliplatin
Form Crystalline solid

RUO – Research Use Only