missing translation for 'onlineSavingsMsg'
Learn More

Oxaliplatin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014909
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014909 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014909 Supplier Merck Emd Millipore Supplier No. PHR1528200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Oxaliplatin, a third generation anti-cancer drug platinum compound, is utilized for the treatment of colorectal cancer. It exhibits a better safety profile than cisplatin (first generation) and carboplatin (second generation).

Specifications

CAS 61825-94-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C8H14N2O4 Pt
MDL Number MFCD00866327
Synonym Oxaliplatin; [SP-4-2-(1 R-trans)]-(1,2-Cyclohexanediamine-N,N′)[ethanedioata(2--)-O,O′]platinμm
Type Certified Reference Material
Quantity 200 mg
Formula Weight 397.29
Grade Certified Reference Material