missing translation for 'onlineSavingsMsg'
Learn More

Oxaliplatin, TCI America™

Catalog No. O0372100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0372100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O0372100MG Supplier TCI America Supplier No. O0372100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61825-94-3
Molecular Formula C8H14N2O4Pt
Molecular Weight (g/mol) 397.29
MDL Number MFCD00866327
InChI Key ZROHGHOFXNOHSO-WHMNLBETNA-L
Synonym (trans-1,2-Cyclohexanediamine)oxalatoplatinum(II)
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine; 1,3-dioxa-2-platinacyclopentane-4,5-dione
SMILES O=C1O[Pt]OC1=O.N[C@@H]1CCCC[C@H]1N

Specifications

Color White
Quantity 100 mg
Formula Weight 397.29
Physical Form Crystalline Powder
Chemical Name or Material Oxaliplatin
RTECSNumber TP2275850
TSCA No
Recommended Storage Refrigerator