missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxaliplatin, 99%

Catalog No. AC456131000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456131000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC456131000 Supplier Thermo Scientific Chemicals Supplier No. 456131000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61825-94-3 | C8H14N2O4Pt | 397.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61825-94-3
Molecular Formula C8H14N2O4Pt
Molecular Weight (g/mol) 397.29
MDL Number MFCD00866327
InChI Key ZROHGHOFXNOHSO-WHMNLBETNA-L
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine; 1,3-dioxa-2-platinacyclopentane-4,5-dione
SMILES O=C1O[Pt]OC1=O.N[C@@H]1CCCC[C@H]1N

Specifications

Melting Point >245.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 2 h)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 100 mg
Formula Weight 397.29
Percent Purity 99%
Chemical Name or Material Oxaliplatin
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Suspected of causing cancer.
May cause allergy or asthma symptoms or breath
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only