CAS RN 6155-57-3
o-Sulfobenzimide Sodium Salt Dihydrate 98.0+%, TCI America™
CAS: 6155-57-3 Molecular Formula: C7H9NNaO5S Molecular Weight (g/mol): 242.201 MDL Number: MFCD00151213 InChI Key: UQKRDMFNUSMUCB-UHFFFAOYSA-N Synonym: saccharin sodium dihydrate,saccharin sodium,sodium saccharin,saccharin soluble,saccharin sodium salt hydrate,1,2-benzisothiazol-3 2h-one, 1,1-dioxide, sodium salt, dihydrate,o-sulfobenzimide sodium salt,2-sulfobenzoic acid imide sodium salt,saccharin sodium salt dihydrate,c7h4nnao3s.2h2o PubChem CID: 46942257 IUPAC Name: 1,1-dioxo-1,2-benzothiazol-3-one;sodium;dihydrate SMILES: C1=CC=C2C(=C1)C(=O)NS2(=O)=O.O.O.[Na]
Sodium Saccharin Dihydrate, Purified, Reagents
CAS: 6155-57-3 Molecular Formula: C7H8NNaO5S Molecular Weight (g/mol): 241.19 InChI Key: NAUHOSQQKJTUAX-UHFFFAOYSA-M IUPAC Name: sodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate dihydrate SMILES: O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
Saccharin Sodium, Dihydrate, Crystalline, USP, 99-101%, Spectrum™ Chemical
CAS: 6155-57-3 Molecular Formula: C7H8NNaO5S Molecular Weight (g/mol): 241.19 InChI Key: NAUHOSQQKJTUAX-UHFFFAOYSA-M IUPAC Name: sodium 1,3-dioxo-3H-1λ6,2-benzothiazol-1-olate dihydrate SMILES: O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
Sodium Saccharin, Dihydrate, Granular, FCC, 98-101%, Spectrum™ Chemical
CAS: 6155-57-3 Molecular Formula: C7H8NNaO5S Molecular Weight (g/mol): 241.19 InChI Key: NAUHOSQQKJTUAX-UHFFFAOYSA-M IUPAC Name: sodium 1,3-dioxo-3H-1λ6,2-benzothiazol-1-olate dihydrate SMILES: O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
Saccharin Sodium, Dihydrate, Powder, USP, 99-101%, Spectrum™ Chemical
CAS: 6155-57-3 Molecular Formula: C7H8NNaO5S Molecular Weight (g/mol): 241.19 InChI Key: NAUHOSQQKJTUAX-UHFFFAOYSA-M IUPAC Name: sodium 1,3-dioxo-3H-1λ6,2-benzothiazol-1-olate dihydrate SMILES: O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
Saccharin Sodium, Dihydrate, Powder, USP, 99-101%, Spectrum™ Chemical
CAS: 6155-57-3 Molecular Formula: C7H8NNaO5S Molecular Weight (g/mol): 241.19 InChI Key: NAUHOSQQKJTUAX-UHFFFAOYSA-M IUPAC Name: sodium 1,3-dioxo-3H-1λ6,2-benzothiazol-1-olate dihydrate SMILES: O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
Sodium Saccharin, Dihydrate, Powder, FCC, 98-101%, Spectrum™ Chemical
CAS: 6155-57-3 Molecular Formula: C7H8NNaO5S Molecular Weight (g/mol): 241.19 InChI Key: NAUHOSQQKJTUAX-UHFFFAOYSA-M IUPAC Name: sodium 1,3-dioxo-3H-1λ6,2-benzothiazol-1-olate dihydrate SMILES: O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12