missing translation for 'onlineSavingsMsg'
Learn More

Sodium Saccharin Dihydrate, Purified, Reagents

Catalog No. 01336978
Change view
Click to view available options
Quantity: 1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01336978 Quantity: 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336978 Supplier Labchem Inc Supplier No. C23247501KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Saccharin Dihydrate, Purified, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6155-57-3
Molecular Formula C7H8NNaO5S
Molecular Weight (g/mol) 241.19
InChI Key NAUHOSQQKJTUAX-UHFFFAOYSA-M
IUPAC Name sodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate dihydrate
SMILES O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Specifications

Boiling Point 439°C
Color White
Physical Form Solid
Quantity 1 kg
Grade Technical
Packaging Poly Bottle
Specific Gravity 2
Chemical Name or Material Sodium Saccharin Dihydrate, Purified