missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saccharin Sodium, Dihydrate, Powder, USP, 99-101%, Spectrum™ Chemical
Description
Spectrum™ Chemical Saccharin Sodium, Dihydrate, Powder, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Saccharin Sodium, Dihydrate, Powder, USP, SO200, C7H4NNaO3S•2H2O
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Poly Bottle |
| Quantity | 25 g |
| Percent Purity | 99 to 101% |
| Grade | USP |
| Physical Form | Crystalline Powder or Lumps |
| Chemical Name or Material | Saccharin Sodium, Dihydrate, Powder |